SP Bel-Art Stepped Tubing Connectors, Bel-Art Products, a part of SP
Catalog #: 16126-396
Supplier: Bel-Art Products, a Part of SP
Specifications
- Tubing I.D.:4.7×6 mm (³⁄₁₆ - ¼")
- Supplier No.:195580000
Specifications
About this item
These fittings ensure a tight, secure connection between tubing having different inner diameters.
- Tight, secure connection
- Polypropylene plastic construction
- Chemically resistant
- Autoclavable
Constructed of chemical-resistant polypropylene plastic for use with a wide range of liquids or gases. FDA-grade for direct contact with food, drugs, and cosmetics. Autoclavable.
Customers Who Bought This Also Bought