Order Entry
ContactUsLinkComponent
VWR* Round Bottles, High-Density Polyethylene, Wide Mouth
VWR® Wide Mouth Bottles, Round, HDPE,
Catalog #: 16125-720
Supplier:  VWR International
Small Business Enterprise
VWR* Round Bottles, High-Density Polyethylene, Wide Mouth
VWR® Wide Mouth Bottles, Round, HDPE,
Catalog #: 16125-720
Supplier:  VWR International
Small Business Enterprise

Specifications

  • Cap Size:
    28-400
  • Capacity:
    30 ml (1 oz.)
  • Cat. no.:
    16125-720

Specifications

About this item

Lightweight bottles are an excellent value for storage of both liquid and powdered process or environmental samples.

  • Lightweight
  • HDPE body construction
  • Polypropylene caps
  • Wide-mouthed design

White polypropylene caps with polyethylene film/foam liners eliminate leaks.

Customers Who Bought This Also Bought