Nalgene® Square Bottles, Polypropylene, Wide Mouth, Thermo Scientific
Catalog #: 16120-962
Supplier: Thermo Fisher Scientific
Specifications
- Cap Size:63-415
- Capacity:1 L (33.8 oz.)
- Color:Translucent
- Cat. no.:16120-962
- Supplier No.:2110-0032
Specifications
About this item
Square, polypropylene wide mouth bottles are designed to replace glass serum and general-purpose storage bottles.
- Resistant to acids, alcohols, and bases
- Contact-clear bottles provide visible liquid level
- Wide mouth for easy cleaning, sample filling, and retrieval
- Square shape saves space and offers large labeling surface
- Leakproof and repeatedly autoclavable
Note: Before autoclaving, set cap on top of bottle without engaging threads.
Ordering information: Supplied with polypropylene screw caps assembled on bottles.
Customers Who Bought This Also Bought