Nalgene® Square Laboratory Bottles, Polypropylene Copolymer, Narrow Mouth, Thermo Scientific
Catalog #: 16120-737
Supplier: Thermo Fisher Scientific
Specifications
- Cap Size:24-415
- Capacity:60 ml (2 oz.)
- Cat. no.:16120-737
- Supplier No.:2016-0060
Specifications
About this item
Square laboratory bottles provide excellent chemical resistance.
- Leakproof
- Repeatedly autoclavable
Note: Before autoclaving, set closure on top of the bottle without engaging the threads.
The translucent design is ideal for long-term storage of inorganic buffers, alcohols, acids, and bases. All sizes have permanent graduations in convenient increments. Bottles are designed for use within a temperature range of 0 to 135 °C (32 to 275 °F).
Ordering information: Supplied with polypropylene screw closures assembled on bottles. For replacement caps for 16120-741 through -753, see 16219-122 series.
Customers Who Bought This Also Bought