Nalgene® Carboys with Handles, Low-Density Polyethylene, Thermo Scientific
Catalog #: 16117-029
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:20 L (5.3 gal.)
- Supplier No.:2210-0050
Specifications
About this item
Useful for storing and containment of chemicals and standards.
- Wide shoulder handles for easy carrying and pouring
- Supplied with ca
- Cap size: 83B
- Accepts No. 13¹/₂ rubber stopper
- Leakproof
Ordering information: For replacement caps, see cap 16225-234 in 16219-122 series.
Customers Who Bought This Also Bought