Order Entry
ContactUsLinkComponent
NALGENE* Rectangular Carboys with Handle, Polypropylene
Nalgene® Rectangular Carboys with Handle, Polypropylene, Thermo Scientific
Catalog #: 16115-147
NALGENE* Rectangular Carboys with Handle, Polypropylene
Nalgene® Rectangular Carboys with Handle, Polypropylene, Thermo Scientific
Catalog #: 16115-147

Specifications

  • Capacity:
    20 L (5.3 gal.)
  • Dimensions:
    31.8 x 21.9 x 38.1 cm (121/2 x 85/8 x 15")
  • Cat. no.:
    16115-147
  • Supplier No.:
    2212-0050

Specifications

About this item

Ideal for storing solutions, media, and sterile water.

  • Leakproof

For replacement caps, see cap 16219-106 in 16219-030 series.

Note: Before autoclaving, set cap on top of carboy without engaging threads.

Useful for handling large volumes of powders and other solid samples. Wide mouth allows easy, efficient cleaning. Space-saving carboys molded in tough, autoclavable polypropylene with 8.9 cm (3¹/₂") neck opening. Sturdy handle attached to molded-in shoulder loops. Calibrated in liters and gallons. Cap size: 100-415.

Customers Who Bought This Also Bought