Order Entry
ContactUsLinkComponent
NALGENE* Boston Round Bottles, Teflon* FEP, Narrow Mouth
Nalgene® Boston Round Bottles, Teflon® Resin FEP, Narrow Mouth, Thermo Scientific
Catalog #: 16071-245
NALGENE* Boston Round Bottles, Teflon* FEP, Narrow Mouth
Nalgene® Boston Round Bottles, Teflon® Resin FEP, Narrow Mouth, Thermo Scientific
Catalog #: 16071-245

Specifications

  • Cap Size:
    38-430
  • Capacity:
    2 L (67.6 oz.)
  • Cat. no.:
    16071-245
  • Supplier No.:
    1600-0064

Specifications

About this item

The most chemical and corrosion-resistant bottle available.

  • Leakproof
  • Autoclavable

Note: Before autoclaving, set cap on top of bottle without engaging threads.

Excellent for high- and low-temperature work, trace metal analysis, and many applications with organic solvents. For high-purity analysis or storage, these bottles can be vigorously cleaned in boiling nitric acid. Bottle with Tefzel® ETFE screw cap withstands temperatures from –105 to 150 °C (–157 to 302 °F).

Packaging: Packaged individually.
Customers Who Bought This Also Bought