Order Entry
ContactUsLinkComponent
NALGENE* Boston Round Bottles, Polypropylene, Narrow Mouth
Nalgene® Boston Round Bottles, Polypropylene Copolymer, Narrow Mouth, Thermo Scientific
Catalog #: 16067-066
NALGENE* Boston Round Bottles, Polypropylene, Narrow Mouth
Nalgene® Boston Round Bottles, Polypropylene Copolymer, Narrow Mouth, Thermo Scientific
Catalog #: 16067-066

Specifications

  • Cap Size:
    24-415
  • Capacity:
    125 ml (4.2 oz.)
  • Cat. no.:
    16067-066
  • Supplier No.:
    2006-0004

Specifications

About this item

Ideal for lab applications.

  • Excellent chemical resistance
  • With polypropylene screw closures
  • Leakproof, Autoclavable

Note: Before autoclaving, set closure on top of bottle without engaging threads.

The 3.8 and 8 L sizes have a built-in shoulder loop for attaching an identification tag.

Ordering information: For replacement closures, see 16219-122 series.
Customers Who Bought This Also Bought