Order Entry
ContactUsLinkComponent
NALGENE* Laboratory Bottles, Polycarbonate, Narrow Mouth
Nalgene® Laboratory Bottles, Polycarbonate, Narrow Mouth, Thermo Scientific
Catalog #: 16058-250
NALGENE* Laboratory Bottles, Polycarbonate, Narrow Mouth
Nalgene® Laboratory Bottles, Polycarbonate, Narrow Mouth, Thermo Scientific
Catalog #: 16058-250

Specifications

  • Cap Size:
    38-430
  • Capacity:
    500 ml (16.9 oz.)
  • Cat. no.:
    16058-250
  • Supplier No.:
    2205-0016

Specifications

About this item

Excellent impact resistance and clarity.

  • 2 and 2.5 L sizes are ideal for use as culture vessels for batch, suspension, or roller culturing
  • 3.8 and 8 L sizes have shoulder loop for attaching identification tags
  • Not recommended for vacuum applications
  • With polypropylene screw closures
  • Leakproof, Autoclavable

Note: Before autoclaving, set closure on top of bottle without engaging threads.

Ordering information: For replacement closures, see 16219-122 series.
Customers Who Bought This Also Bought