VWR® Disposable Pasteur Pipettes, Borosilicate Glass 7.0 Expansion
Catalog #: 14672-380
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Description:Disposable Pasteur Pipet
- Cat. no.:14672-380
- Overall Length:22.9 cm (9")
Specifications
About this item
VWR® Disposable Pasteur Pipettes are ideally suited for use in both clinical and industrial laboratories.
- Made up of borosilicate glass 7.0
- Ideal for both clinical and industrial use
- Supplied in convenient USE-Packs® packaging
Disposable Pasteur pipettes provide a convenient and time-efficient means for inoculating culture media and transferring samples and reagents in both clinical and industrial laboratories. No cotton plug. Pipette body Ext.Ø: 7 mm. Capillary stem Ext.Ø: approximately 1.5 mm. Controlled drop pipette (14672-608) delivers 22 to 28 drops per ml.
Packaging: Supplied in convenient USE-Packs® packages, each containing 200 pipettes.
Customers Who Bought This Also Bought