Samco™ Disposable Transfer Pipettes, Fine Tip, Thermo Scientific
Catalog #: 14670-329
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:1.5 ml
- Sterility:Sterile
- Description:Extended Fine Tip, Small Bulb
- Cat. no.:14670-329
- Packaging:20/Bag
- Overall Length:10.4 cm (4")
- Stem Diameter:3.0 mm
- Bulb Draw:1.0 ml
- Supplier No.:231-20S
Specifications
About this item
These fine-tip polyethylene transfer pipettes are excellent for the precise dispensing of small drops (45 to 50 per ml) into microbiological Int.Ø.
Strips, ELISA plates, and microcentrifuge and precipitation tubes. Ideal for removing cell layers, filling electrophoresis wells, spotting chromatography plates, and other similar applications.
Customers Who Bought This Also Bought