Samco™ Disposable Transfer Pipettes, Graduated, Thermo Scientific
Catalog #: 14670-205
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:5.8 ml
- Sterility:Non sterile
- Description:Large Bulb
- Packaging:Bulk Packed
- Overall Length:15.5 cm (6⅛")
- Stem Diameter:4.6 mm
- Graduations:1.0 mL
- Bulb Draw:3.4 ml
- Supplier No.:222
- Drops per mL:25
Specifications
About this item
Graduated polyethylene transfer pipettes are excellent for sampling of serum, plasma, blood, urine, and other biological fluids, especially where more accurate volume measurements are required.
Sterile models are specially packaged for batch-testing applications in microbiology and bacteriology labs. Nonsterile models are suitable for single, specialized research type testing. Stem narrows down to a fine tip.
Customers Who Bought This Also Bought