VWR® Autoclavable Bags, Nonhazardous Waste
Catalog #: 14220-040
Supplier: VWR International
Small Business Enterprise
Specifications
- W×L:483×584 mm (19×23")
- Color:Clear
Specifications
About this item
Intended for use in autoclaving nonhazardous waste and constructed of high-integrity polypropylene, these bags can withstand autoclaving temperatures up to 135 °C (275 °F).
- Superior tear resistance
- Welded seals
Heavy walls provide superior tear resistance and welded seals minimize risk of leakage. Bags meet the strength requirements of the 165g dart drop standard performed per ASTM D1709-98. All bags are clear.
Certifications: Meets the 165 g dart drop standard (ASTM D1709-98).
Customers Who Bought This Also Bought