Order Entry
ContactUsLinkComponent
VWR* Autoclavable Biohazard Bags
VWR® Autoclavable Biohazard Bags and Containers
Catalog #: 14220-034
Supplier:  VWR International
Product available on GSA Advantage®
Small Business Enterprise
VWR* Autoclavable Biohazard Bags
VWR® Autoclavable Biohazard Bags and Containers
Catalog #: 14220-034
Supplier:  VWR International
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • W×L:
    914×1143 mm (36×45")
  • Thickness:
    2 mil
  • Cat. no.:
    14220-034

Specifications

About this item

Double-thick bags are ideal for sterilizing animal cages and other objects with sharp corners.

  • High-integrity polypropylene
  • Available with temperature indicator
  • Resists punctures, tears, and leaks

Ideal for processing biohazardous materials that require autoclaving prior to disposal.

Twist-tie closure allows bag to expand and vent steam during cooling. Bags resist punctures, tears, and leaks and meet the 165g dart drop standard performed as required by ASTM D1709-98. Each bag is imprinted with the universal biohazard symbol.

Indicator bags have the words 'Contents Have Been Autoclaved' printed in a chemically active ink. Before autoclaving, the ink is white. After autoclaving is complete, it darkens, providing instant proof that biohazardous waste has been autoclaved.

Biohazard bag containers are made of rigid, high-density polyethylene for durability and leak-free performance. Snap-on, snap-off lid provides an extra measure of safety in the lab. White with universal biohazard waste symbol imprinted in black on an orange background. Containers are not autoclavable.

Certifications: Meets the 165 g dart drop standard (ASTM D1709-98).
Customers Who Bought This Also Bought