WHEATON® ABC Screw Caps, DWK Life Sciences
Catalog #: 14213-300
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Cap Size:9 mm
- Septum:Red PTFE/White Silicone
- Description:ABC Cap
- Material:PP
- Cap Color:Natural
- Supplier No.:W225330-01
Specifications
About this item
These closures for ABC Vials feature Wheaton Super Septa and low extractable septa.
- Manufactured from polypropylene
- 100/pack, 10 packs/case
Available in natural, black, red, blue, yellow or green polypropylene
Customers Who Bought This Also Bought