To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Ø:12 mm
- Accuracy:±0.02 pH
- Length:110 mm
- Material:Epoxy body
- Junction:Open
- Operating temperature range:0 to 60°C
- Reference system:Internal Ag/AgCl Double Junction
- Cat. no.:14004-266
- Cable Length:1 m
- Connector:BNC, waterproof and 8-pin Mini-DIN
- Supplier No.:9107APMD
Specifications
About this item
Read viscous liquid samples quickly and accurately without the sample coming into contact with silver ions with Orion™ AquaPro™ pH Combination Electrodes.
- Always store in Orion™ pH Electrode Storage Solution
- Either store in a beaker of storage solution or reassemble in the provided electrode storage bottle
- pH range is 0 to 14
Orion™ AquaPro™ pH Combination Electrodes have a clog-resistant and open double junction reference system. These electrodes are ideal for measurements in samples containing proteins or TRIS buffers, which do not contain mercury. The electrodes require minimal maintenance because there is no fill solution; instead they have a polymer gel reference.