Order Entry
ContactUsLinkComponent
STANDARD CNDTV 111.9 MS/CM 15ML PK10
ORION® Conductivity Standards, Thermo Scientific
Catalog #: 14002-862
CAS Number:  
STANDARD CNDTV 111.9 MS/CM 15ML PK10
ORION® Conductivity Standards, Thermo Scientific
Catalog #: 14002-862
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Specific Conductance: 111.9 mS/cm
  • Accuracy:
    ±1% for 2 years
  • Supplier No.:
    1100510

Specifications

About this item

These ready-to-use standards allow five conductivity levels to be easily measured in the field

Standards come in convenient 15mL pouches and certified against standards whose calibrations are traceable to the National Institute of Standards and Technology.

Ordering information: Store at room temperature.
Customers Who Bought This Also Bought