Order Entry
ContactUsLinkComponent
NALGENE* Graduated Transparent Beaker with Handle, PMP
Nalgene® Graduated Transparent Beakers with Handle, PMP, Thermo Scientific
Catalog #: 13916-999
NALGENE* Graduated Transparent Beaker with Handle, PMP
Nalgene® Graduated Transparent Beakers with Handle, PMP, Thermo Scientific
Catalog #: 13916-999

Specifications

  • Capacity:
    3 L (101.4 oz.)
  • Graduation range:
    200 - 3000 ml
  • Cat. no.:
    13916-999
  • Supplier No.:
    1223-3000

Specifications

About this item

Polymethylpentene (PMP) material allows a clear view of beaker contents.

  • Rectangular handle for easy carrying and pouring
  • Molded-in graduations in both mL and oz. will not wear off with continuous use or washings

Excellent chemical resistance to acids, bases and aliphatic alcohols and useful for a broad range of solutions. Withstand continuous exposure to temperatures as high as 150°C (302°F) and intermittent temperatures to 175°C (347°F).

Customers Who Bought This Also Bought