Order Entry
ContactUsLinkComponent
NALGENE* Graduated Beaker with Handle, HDPE
Nalgene® Graduated Beakers with Handle, High-Density Polyethylene, Thermo Scientific
Catalog #: 13916-977
NALGENE* Graduated Beaker with Handle, HDPE
Nalgene® Graduated Beakers with Handle, High-Density Polyethylene, Thermo Scientific
Catalog #: 13916-977

Specifications

  • Capacity:
    3 L (101.4 oz.)
  • Graduation range:
    200 - 3000 ml
  • Cat. no.:
    13916-977
  • Supplier No.:
    1220-2500

Specifications

About this item

Made from high-density polyethylene with extra heavy wall

  • Graduated in both ounces and milliliters
  • Large rectangular handle

Especially useful in handling acids, alkalies, and photographic chemicals.

Customers Who Bought This Also Bought