Order Entry
ContactUsLinkComponent
Graduated Beaker with Handle, Low Form, Polypropylene
VWR®, Graduated Beakers with Handle, Low Form, Polypropylene
Catalog #: 13916-730
Supplier:  VWR International
Graduated Beaker with Handle, Low Form, Polypropylene
VWR®, Graduated Beakers with Handle, Low Form, Polypropylene
Catalog #: 13916-730
Supplier:  VWR International

Specifications

  • Capacity:
    1 L (34 oz.)
  • Graduation range:
    50 - 1000 ml
  • Cat. no.:
    13916-730

Specifications

About this item

Graduated beakers are robust, with a non-drip spout and ridged thumb-grip on handle.

  • Molded of autoclavable, translucent polypropylene
  • Low form for better stability
  • All sizes graduated in milliliters and liters
Customers Who Bought This Also Bought