Order Entry
ContactUsLinkComponent
NALGENE* Polypropylene Griffin Low Form Graduated Beakers
Nalgene® Griffin Low Form Graduated Beakers, Polypropylene, Thermo Scientific
Catalog #: 13915-114
NALGENE* Polypropylene Griffin Low Form Graduated Beakers
Nalgene® Griffin Low Form Graduated Beakers, Polypropylene, Thermo Scientific
Catalog #: 13915-114

Specifications

  • Capacity:
    400 ml (14 oz.)
  • Graduation range:
    50 - 400 ml
  • Cat. no.:
    13915-114
  • Supplier No.:
    1201-0400

Specifications

About this item

Heavy-duty, translucent polypropylene beakers with ribs for stacking.

  • Easy-to-read graduations
  • Autoclavable
  • Chemically resistant

Beakers feature easy-to-read graduations with size and resin codes. Maximum use temperature and no-flame symbol printed in black. Autoclavable and chemically resistant. Not for use on a hot plate.

Customers Who Bought This Also Bought