Order Entry
ContactUsLinkComponent
VWR* Griffin Beakers, Polypropylene
VWR®, Griffin Beakers, Polypropylene
Catalog #: 13890-160
Supplier:  VWR International
VWR* Griffin Beakers, Polypropylene
VWR®, Griffin Beakers, Polypropylene
Catalog #: 13890-160
Supplier:  VWR International

Specifications

  • Capacity:
    2 L (68 oz.)
  • Graduation range:
    200 - 2000 ml
  • Cat. no.:
    13890-160

Specifications

About this item

Translucent walls and molded graduations make it easy to prepare, mix, and measure solutions within the beaker.

  • Stackable
  • Sturdy construction withstands repeated autoclaving
  • Break-resistant, non-toxic, non-contaminating, and extremely chemical-resistant
  • Feature no-drip spout for clean pouring
  • Tapered walls for secure grip
  • Oval marking spot

Ideal beaker for general laboratory procedures.

Customers Who Bought This Also Bought