Order Entry
ContactUsLinkComponent
Antistatic Weighing Canoe, Medium
Antistatic Weighing Canoes, Dyn-A-Med Products
Catalog #: 13451-989
Supplier:  Dyn-A-Med
Antistatic Weighing Canoe, Medium
Antistatic Weighing Canoes, Dyn-A-Med Products
Catalog #: 13451-989
Supplier:  Dyn-A-Med

Specifications

  • Color:
    Natural
  • Material:
    Polystyrene
  • Size:
    Medium
  • Cat. no.:
    13451-989
  • Interior Dimensions:
    7.3W x 12.1L x 3.2D cm (2 7/8 x 4 3/4 x 1 1/4")
  • Supplier No.:
    80056

Specifications

About this item

Anti-static Weighing Canoes are designed for accurate weighing of static-affected material.

  • Disposable
  • Polystyrene construction
  • Anti-static finish

Weighing Canoes are made of recyclable virgin polystyrene with 109 ohm2 resistivity. Contoured sides provide a convenient non-slip gripping surface. Canoe shape allows easy pouring of weighed samples. Available in three sizes.

Customers Who Bought This Also Bought