ContactUsLinkComponent
Capillary Tubes
Capillary Tubes, Drummond
Catalog # 13450-216
Supplier:  Drummond
undefined
Capillary Tubes, Drummond
Catalog # 13450-216
Supplier:  Drummond
Supplier Number:  1-000-800/12
Specifications
  • Description:
    Capillary Tubes
  • Cat. no.:
    13450-216
  • Supplier No.:
    1-000-800/12
Specifications
About this item
Glass capillary tubes for coagulation, hemoglobin, prothrombin, blood grouping and typing.
  • Capacity 9.4 µL
  • Length 7.5cm
  • Internal Diameter 0.4-0.6mm.
Ordering information: Packaged in vials of 100 tubes.
Frequently Bought Together