VWR® Bath Fluids
Catalog #: 13272-034
Supplier: VWR International
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:PolyScience polycool HC -50, Freezing Point –56 °C
- Size:3.8 l (1 gal.)
- Temperature Range:–50...+100 °C (–58...+212 °F)
- For Use With:VWR Refrigerated Circulating Baths
- Viscosity:(0°C) 3.6 cSt
(–40°C) 15.1 cSt
Specifications
About this item
These heat-transfer fluids provide superior thermal stability for high or low temperatures.
- Silicone heat transfer fluid
Check compatibility of fluids before using, as fluids are incompatible with Buna or natural rubber.
Packaging: Bottles are packaged in a carton for shipment.
Customers Who Bought This Also Bought