Order Entry
ContactUsLinkComponent
VIKEM VINYL MICROSCOPE COVER
SP Bel-Art Vikem® Microscope Covers, Vinyl, Bel-Art Products, a part of SP
Catalog #: 13197-180
VIKEM VINYL MICROSCOPE COVER
SP Bel-Art Vikem® Microscope Covers, Vinyl, Bel-Art Products, a part of SP
Catalog #: 13197-180

Specifications

  • Ø:
    330 mm (13") (bottom); 230 mm (9") (top)
  • Description:
    Vikem® vinyl microscope cover, standard size
  • Height:
    420 mm (16.5")
  • Cat. no.:
    13197-180
  • Supplier No.:
    243010000

Specifications

About this item

Transparent Vikem® vinyl covers protect valuable microscopes, balances, and other instruments from dust and spills.

  • Transparent
  • Easy application

Simply slide the cover over the instrument when it is not in use.

Ordering information: Available in three standard sizes.
Customers Who Bought This Also Bought