VWR® Weighing Paper
Catalog #: 12578-201
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Size:15.2×15.2 cm (6×6")
Specifications
About this item
Weighing paper is designed for analytical weighing with minimal influence on analytical results for weighing and transporting sample.
- Clean and dust-free
- Lightweight
- Nitrogen-free
- Smooth surface
A smooth surface is ideal for transferring weighed material. Clean, dust-free sheets. Paper is lightweight, yet strong enough to support analytical size samples.
Packaging: All sizes of weighing papers are packaged 500 sheets per box.
Customers Who Bought This Also Bought