Order Entry
ContactUsLinkComponent
Glass Fiber Pads
Glass Fiber Pads for Moisture Analyzers, Dyn-a-Med
Catalog #: 12578-105
Supplier:  Dyn-A-Med
Glass Fiber Pads
Glass Fiber Pads for Moisture Analyzers, Dyn-a-Med
Catalog #: 12578-105
Supplier:  Dyn-A-Med

Specifications

  • Pan/Platform Size:
    127 x 127 mm
  • Description:
    Round, 9 cm (31/2") Diameter
  • Cat. no.:
    12578-105
  • Supplier No.:
    80082

Specifications

About this item

Glass fiber pads conveniently hold liquid or semi-solid samples for moisture determination

For use in microwave or infrared moisture analyzers. Also may be used in furnaces. 100% borosilicate. Available as square or round pads.

Customers Who Bought This Also Bought