Order Entry
ContactUsLinkComponent
Bitran* Liquid Tight Specimen Bags
Bitran® Liquid-Tight Specimen Bags, Com-Pac
Catalog #: 11217-126
Supplier:  Com-Pac
Bitran* Liquid Tight Specimen Bags
Bitran® Liquid-Tight Specimen Bags, Com-Pac
Catalog #: 11217-126
Supplier:  Com-Pac

Specifications

  • W×L:
    230×301 mm (9×12")
  • Cat. no.:
    11217-126
  • Supplier No.:
    4755-PE

Specifications

About this item

Polyethylene and SARANEX® zipper bags are designed for the handling and storage of specimens.

  • Unique manufacturing process eliminates leaks
  • Double track airtight zipper is easy to close

The SARANEX® (Series S) bags provide an exceptional vapor barrier for optimum odor control. Polyethylene (Series PE) bags remain flexible when frozen during storage.

Customers Who Bought This Also Bought