SP Bel-Art Bench-Top Biohazard Bags, Bel-Art Products, a part of SP
Catalog #: 11215-857
Supplier: Bel-Art Products, a Part of SP
Specifications
- Capacity:1,6 L (0.43 gal.)
- W×L:220×280 mm (8.5×11")
- Description:Biohazard bags
- Thickness:0.018 mm (0.72 mil)
- Cat. no.:11215-857
- Supplier No.:F13166-0000
Specifications
About this item
Bright red, autoclavable bags feature one long flap for easy opening. Printed with the biohazard symbol and sterilization indicator patch, they are made of High Molecular Weight, High-Density (HMHD) polyethylene.
Durable cover provides compliance with 29 CFR 1910.1030(d)(2)(xiii)(A), which requires a cover on any container used to store, transport, or ship biohazardous wastes.
- Use as a biohazard material handler: Small size allows the bag to be inverted over your hand to safely pick up waste
- Steam autoclavable at 121 °C (250 °F)
Bright red, autoclavable bags feature one long flap for easy opening. Printed with the biohazard symbol and sterilization indicator patch, they are made of High Molecular Weight, High-Density (HMHD) polyethylene.
For a space-saving disposal unit in fume hoods or on bench tops, use with Poxygrid® Benchtop Biohazard Bag Holder (11215-508) and Cover (89237-571).
Certifications: Pass ASTM 1922 tear resistance and ASTM 1709 dart impact tests.
Customers Who Bought This Also Bought