VWR® Autoclavable PE Biohazard Bags, 4 mil
Catalog #: 11215-822
Supplier: VWR International
Small Business Enterprise
Specifications
- W×L:304×610 mm (12×24")
- Color:red
- Cat. no.:11215-822
Specifications
About this item
These heavy-duty bags with extra puncture resistance and high impact strength are constructed of high-molecular, high-density 4 mm polyethylene plastic.
- Bright red color for quick identification
- Clearly labeled with biohazard warnings
Autoclavable up to 121 °C (250 °F). Ideal for sterilizing animal cages. Each package includes 20.3 cm (8") twist ties. Bags meet many state and local regulations.
Customers Who Bought This Also Bought