VWR® Biohazard Specimen Bags
Catalog #: 11215-684
Supplier: VWR International
Small Business Enterprise
Specifications
- Dimensions:15.2×22.9 cm (6×9")
- Cat. no.:11215-684
Specifications
About this item
Designed for safe and secure transport of a specimen container and documentation, these 2 mm bags feature a pouch to keep the paperwork separate from the container.
- Safe and secure specimen transport
- Store paperwork separate from biohazard specimen bag
Convenient transport pouch for specimen container and documentation.
Customers Who Bought This Also Bought