Axygen® Pipette Filter Tip, Axygen®, Corning
Catalog #: 10011-884
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Volume:200 µl
- Cat. no.:10011-884
- Sterile:-
- Supplier No.:TF-200
Specifications
About this item
Axygen brand filter tips allow free passage of air while preventing passage of aerosols.
The filters contain no leachable additives.
Customers Who Bought This Also Bought