ContactUsLinkComponent
Conical-Bottom Screw Cap Microtube with Cap
Axygen® Screw Top Microcentrifuge Tubes with Caps, Polypropylene, Corning
Catalog #: 10011-598
Supplier:  Corning
Product available on GSA Advantage®
Conical-Bottom Screw Cap Microtube with Cap
Axygen® Screw Top Microcentrifuge Tubes with Caps, Polypropylene, Corning
Catalog #: 10011-598
Supplier:  Corning
Product available on GSA Advantage®
Specifications
  • Sterility:
    Non sterile
  • Volume:
    2 ml
  • Maximum RCF:
    20000×g
  • Cap Color:
    Clear
  • Supplier No.:
    SCT-200-C
Specifications
About this item
These conical-bottom screw cap microcentrifuge tubes serve a broad range of laboratory needs including collection, pressing, centrifugation, long-term storage, and OEM packaging and shipping of restriction enzymes or other DNA/RNA modifying reagents.
These conical-bottom screw cap microcentrifuge tubes serve a broad range of laboratory needs including collection, pressing, centrifugation, long-term storage, and OEM packaging and shipping of restriction enzymes or other DNA/RNA modifying reagents.
Color-coded caps provide a syringe port that is easily pierceable for radioisotope labeling and fraction collecting. The threaded O-ring design ensures a uniform closure allowing centrifugation up to 20000×g. Microcentrifuge tubes have ribs on the cap and tube to fit the Axyrack™ Microtube Racks to allow single-handed opening and closing. Polypropylene tubes are certified RNase-, DNase-, and pyrogen-free.
Ordering information: Tubes without ribs are also available; contact your sales representative for information.
All microtubes include non-looped caps with O-rings. Presterilized microtubes are supplied with cap attached.
Customers Who Bought This Also Bought