Axygen® Pipette Filter Tip, Axygen®, Corning
Catalog #: 10011-348
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Volume:1000 µl
- Sterile:-
- Supplier No.:TF-1000
Specifications
About this item
Axygen brand filter tips allow free passage of air while preventing passage of aerosols.
The filters contain no leachable additives.
Customers Who Bought This Also Bought