Axygen® Research-Grade Pipette Tips, Corning
Catalog #: 10011-306
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Volume:1000 µl
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Packed:Bulk
- Supplier No.:T-1000-C
Specifications
About this item
These tips are designed to fit a wide variety of 1000 µl single-channel pipettors.
Flexible walls and a series of internal sealing rings ensure a secure fit and less force required to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry. Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures.
Customers Who Bought This Also Bought