Axygen® Automation-Compatible PCR Sealing Mats, Corning
Catalog #: 10011-130
Supplier: Corning
Specifications
- Description:AxyMat™ Sealing Mat
- Cat. no.:10011-130
- Supplier No.:AM-96-PCR-RD
Specifications
About this item
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications.
- Minimize evaporation
- Made of research-grade silicone
- Autoclavable
They can be reused following a bleach wash and ethanol rinse protocol.
Customers Who Bought This Also Bought