VWR® Atlas Shelf Brackets
Catalog #: VWRBLC36
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Depth:36"
- Color:Light neutral
- Description:Low-Profile Center-Mounted Shelf Brackets
- Cat. no.:VWRBLC36
Specifications
About this item
Designed to be infinitely adjustable along vertical Atlas uprights.
- Brackets are powder-coated with high-quality chemical-resistant epoxy powder
- Constructed of 12-gauge steel
Shelf brackets provide a sturdy base of support for shelving units and are adjustable along vertical Atlas uprights.
Ordering information: Brackets are sold in sets of two. Low-profile shelf brackets and Kableway shelf brackets are designed for under mount only. For Atlas uprights, see VWRWMUP24, VWRC24, and VWRSU24 series. For Atlas shelving, see VWRNL1236, VWR1L1236, and VWR2L1236 series.
Customers Who Bought This Also Bought