Order Entry
ContactUsLinkComponent
Bovie Serum Albumi
Bovine Serum Albumin (BSA)
Catalog #: RLBSA-1000
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Bovine Serum Albumin (BSA)
Catalog #: RLBSA-1000
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Protease-free:
    Y
  • Source:
    Serum
  • Species:
    Bovine
  • Size:
    1 kg
  • Protein synonyms:
    BSA Blocker|BSA|FACS Buffer|BSA Blocking|Fraction V|Cohn Fraction V|Albumin
  • Protein/peptide name:
    Bovine Serum Albumin
  • pH:
    pH 7
  • Supplier No.:
    BSA-1000

Specifications

About this item

This product is suitable for use in protease sensitive assays such as RIA, EIA and nucleic acid hybridization, use as a stabilizing agent for proteins and enzymes including dilute solutions of antibody, use as a blocking agent to reduce non-specific binding.

  • IgG-free
  • Protease-free

This product is suitable for use in protease sensitive assays such as RIA, EIA and nucleic acid hybridization, use as a stabilizing agent for proteins and enzymes including dilute solutions of antibody, use as a blocking agent to reduce non-specific binding.

Customers Who Bought This Also Bought