Phenolphthalein 1% (w/v) in 95% (v/v) alcohol indicator, neutralized
Catalog #: RC562032
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number: 77-09-8
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle
- Size:1 L
- Cat. no.:RC562032
- Supplier No.:R5620000-1A
Specifications
Specification Test Results
- Appearance:Clear, very light pink, alcohol odor
- Suitability for Use:Colorless (pH 8.0) - Pink (pH 10.0)
- Phenolphthalein 1% (w/v) in 95% (v/v) alcohol indicator, neutralized:
Specification Test Results
About this item
Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
Formula:C₂₀H₁₄O₄
Boiling Pt:∼78 °C (1013 hPa)
Density:∼0.85 g/cm³ (20 °C)
MDL Number:MFCD00005913
CAS Number:77-09-8
:Storage Temperature: Ambient
ADR:3,II
:Length
:Width
:Height
:Diameter
:Thickness
:Boiling Pt:
:Boiling Pt: ? 78 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 258 to 262 °C
:Flash Pt:
:Flash Pt:
:M.W. 318.33 g/mol
:Phenolphthalein (1 - 5%) in ethanol
Storage Temperature:Ambient
EINECS:201-004-7
:? 78 °C (1013 hPa)
UN:1170
:3
:II
:GHS02
:GHS08
:12,07392
:14,07243
Customers Who Bought This Also Bought