Order Entry
ContactUsLinkComponent
Conductivity/TDS Standard, 4L
Conductivity Standard, 10 µS/cm (10 µmho/cm) at 25°C 4.7 ppm as NaCl
Catalog #: RC22461
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
Conductivity/TDS Standard, 4L
Conductivity Standard, 10 µS/cm (10 µmho/cm) at 25°C 4.7 ppm as NaCl
Catalog #: RC22461
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Conductivity:
    10000 µmhos/cm
  • Size:
    3.8 L (1 gal.)
  • Cat. no.:
    RC22461
  • TDS:
    5400 ppm NaCl
  • Supplier No.:
    R2246000-4A

Specifications

About this item

Calibration standards are individually calibrated and certified directly traceable to National Institute of Standards and Technology Electrolytic Conductivity Standard Reference Materials.

Contact your VWR representative for custom-prepared conductivity standards.

Each plastic container is labeled with the certified conductivity value, TDS concentration expressed as ppm sodium chloride (NaCl), and TDS expressed as equivalent 'Fresh Water Ion' concentrations when appropriate.

Customers Who Bought This Also Bought