Conductivity Standard, 10 µS/cm (10 µmho/cm) at 25°C 4.7 ppm as NaCl
Catalog # RC22431
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Size:3.8 L (1 gal.)
- Cat. no.:RC22431
- Conductivity:1000 µmhos/cm
- TDS:495 ppm NaCl
- Supplier No.:R2243000-4A
Specifications
About this item
Calibration standards are individually calibrated and certified directly traceable to National Institute of Standards and Technology Electrolytic Conductivity Standard Reference Materials.
Contact your Avantor representative for custom-prepared conductivity standards.
Each plastic container is labeled with the certified conductivity value, TDS concentration expressed as ppm sodium chloride (NaCl), and TDS expressed as equivalent 'Fresh Water Ion' concentrations when appropriate.
Frequently Bought Together