Silicone Stoppers with Holes, Kimble Chase
Catalog #: KT953715-0801
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Cat. no.:KT953715-0801
- Stopper No.:8
- Hole Size:9.5 mm (3/8")
- Supplier No.:953715-0801
Specifications
About this item
General-purpose, single-hole silicone rubber stoppers
Steam autoclavable. Opaque yellowish-white color. Stoppers are manufactured from pure silicone rubber.
Customers Who Bought This Also Bought