KIMBLE® Natural Rubber Septa for NMR Tubes, DWK Life Sciences
Catalog #: KT8970980010
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Ø:10 mm
- Color:Red
- Description:Rubber septa for NMR tubes
- Cat. no.:KT8970980010
- Supplier No.:897098-0010
Specifications
About this item
Soft, natural rubber allows easy penetration.
- Inner and outer seal
- Ideal for thin-walled tubes
- Penetration point for syringe or cannulation additions
Useful as a closure or for sample outgassing.
Customers Who Bought This Also Bought