Accessories for KIMBLE® MICROFLEX® Williamson Microscale Kits, DWK Life Sciences
Catalog #: KT748017-0020
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Description:Polyethylene disc, 20 Microns
- Cat. no.:KT748017-0020
- Supplier No.:748017-0020
Specifications
About this item
Test fabric is useful for testing dyes on a variety of fabrics
Three-foot length of fabric four inches wide features 8 strips of different fibers woven side by side. Filament acetate, bleached cotton, spun polyamide type 66, polyester disperse dyeable, spun polyacrylic, spun silk, spun viscose, and worsted wool.
Accessory for Microscale Williamson kit.
Customers Who Bought This Also Bought