One-Way Stopcock Valves, Female to Male, Kimble Chase
Catalog #: KT420163-0000
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Color:Blue
- Description:One-way stopcock valve, polycarbonate body, HDPE plug, female luer to male luer
- Supplier No.:420163-0000
Specifications
About this item
One-way stopcocks with a female to male connection and HDPE plug.
Customers Who Bought This Also Bought