Specifications
- Description:Developing tank with nonslip lid
- Cat. no.:KT416185-0000
- W×D×H:27×7×25 cm (105/8×23/4×97/8")
- Supplier No.:416185-0000
Specifications
About this item
Sturdy molded glass developing tanks feature clear sides to allow unobstructed visual inspection of TLC plates up to 20x20cm (77/8x77/8") in size
Tanks are available with a standard, nonslip, or latch style lid. The nonslip lid has a knob for easy lifting, and PTFE retainers inside prevent the lid from sliding off the tank. The latch lid holds lid firmly to the tank, creating a stable environment for developing TLC plates. The latch lid also has a knob for easy lifting and PTFE retainers inside.
Nonslip and latch tank lids are available with holes drilled for #0 rubber stoppers to facilitate the addition of reagents with minimal disruption to the environment of developing plates.
Edges are beveled to eliminate sharp edges. Tanks also have a ground bottom and top for a level surface. A raised edge along the inside bottom allows the simultaneous development of two 20x20cm (77/8x77/8") or four 5x20cm (2x77/8") plates.
Weight: 5.4 kg (11.9 lbs.)