KONTES® Reaction Beaker, Glass Jacketed, Kimble Chase
Catalog #: KT317000-0100
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Capacity:100 ml (3.4 oz.)
- O.D.:80 mm (Outside), 47 mm (Inside)
- Height:85 mm (Outside), 73 mm (Inside)
- Supplier No.:317000-0100
Specifications
About this item
Useful in biological and chemical reactions including titrations, enzyme digestion and synthetic organic reactions.
- Jacket has #2 inlet/outlet serrated hose connections
- Conical beaker bottom accepts egg-shaped PTFE magnetic stir bar for optimum agitation without spin-out
- Open top provides good access to concentrated residual samples
Borosilicate glass construction designed for temperature controlled reactions.
Certifications: Design meets ASTM Specification E960, Type I requirements. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought