Open-Top Septa Caps, Thermo Scientific
Catalog #: IR297-2400UB
Supplier: Thermo Scientific
UL Listed
Specifications
- Cap Size:24-400
- Color:White
- Description:Open-Top Septa Cap
- Material:PP
- Liner Material:Bonded 0.060" PTFE-lined Silicone Septa
- Supplier No.:297-2400UB
Specifications
About this item
These Open-Top Septa Caps are available with or without a bonded-in PTFE-lined silicone septa
They are the same high-quality polypropylene septa caps used on Thermo Scientific VOA vials, septa bottles, and septa jars. Caps are processed to meet EPA performance-based specifications for Volatile Organic Analysis.
Customers Who Bought This Also Bought