Loose Septa for 24-414 Open-Top Caps, Thermo Scientific
Catalog #: IR288-0022BP
Supplier: Thermo Scientific
Specifications
- Color:Natural
- Description:Bonded PTFE-lined silicone septa, 2000
- Thickness:0.125"
- Cat. no.:IR288-0022BP
- Supplier No.:288-0022BP
Specifications
About this item
Loose Septa are designed to serve as replacement septa for Thermo Scientific VOA vials and septa bottles with 24-414 open-top caps.
Loose septa may also be used as replacement parts for VOA vials and bottles with unbonded septa.
Certifications: Processed to meet EPA Performance Based Specifications for Volatile Organic Analysis.
Ordering information: Caps may also be purchased separately for use with this septum.
Customers Who Bought This Also Bought